Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144844.2 | NP_659093.2 | 6 | 459 | ||
GenBank mRNA | AK148002.1 | - | 6 | 449 | ||
AK185546.1 | - | 2 | 128 | |||
AY046947.1 | - | 6 | 476 | |||
BC006915.1 | - | 6 | 456 | 3601148 | ||
BC049802.1 | - | 6 | 425 | 6332730 |