Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144868.3 | NP_659117.2 | 2 | 1028 | ||
GenBank mRNA | AF237953.1 | - | 2 | 269 | ||
AK171516.1 | - | 2 | 815 | |||
BC054751.1 | - | 2 | 307 | |||
BC055289.1 | - | 3 | 831 | |||
BC096538.1 | - | 2 | 1028 |