Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144887.4 | NP_659136.1 | 3 | 1353 | ||
GenBank mRNA | AK082513.1 | - | 3 | 1278 | ||
AK145086.1 | - | 3 | 1280 | |||
AK173251.1 | - | 3 | 1308 | |||
AY894891.1 | - | 3 | 1345 | |||
BC020051.1 | - | 2 | 323 | 3987696 | ||
BC065155.1 | - | 3 | 1223 | 5706087 |