Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144844.2 | NP_659093.2 | 7 | 599 | ||
GenBank mRNA | AK148002.1 | - | 7 | 589 | ||
AY046947.1 | - | 7 | 616 | |||
BC006915.1 | - | 7 | 596 | 3601148 | ||
BC049802.1 | - | 7 | 565 | 6332730 |