Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144868.3 | NP_659117.2 | 8 | 2744 | ||
GenBank mRNA | AF237953.1 | - | 6 | 761 | ||
AK171516.1 | - | 7 | 2981 | |||
BC025838.1 | - | 2 | 277 | |||
BC038056.1 | - | 3 | 852 | |||
BC054751.1 | - | 8 | 2023 | |||
BC055289.1 | - | 9 | 2547 | |||
BC096538.1 | - | 8 | 2744 |