Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033597.1 | NP_001028769.1 | 19, 20 | 2048 | ||
NM_001033598.1 | NP_001028770.1 | 19, 20 | 2017 | |||
NM_001033599.1 | NP_001028771.1 | 19, 20 | 2223 | |||
NM_144823.4 | NP_659072.3 | 19, 20 | 2048 | |||
GenBank mRNA | AB073974.1 | - | 18, 19 | 1892 | ||
AK122384.1 | - | 8, 9 | 3814 | |||
AK147283.1 | - | 19, 20 | 2018 | |||
AK147421.1 | - | 19, 20 | 2049 | |||
AY167035.1 | - | 19, 20 | 2018 | |||
AY786360.1 | - | 21, 22 | 2566 | |||
AY786361.1 | - | 19, 20 | 1998 | |||
AY786362.1 | - | 19, 20 | 2190 | |||
AY786363.1 | - | 19, 20 | 2043 | |||
BC016114.1 | - | 19, 20 | 1974 | 4481949 | ||
BC022959.1 | - | 19, 20 | 2048 | 5362660 |