Search Thermo Fisher Scientific
Gene Symbol: Rab29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144875.2 | NP_659124.2 | 6 | 808 | ||
GenBank mRNA | AB232625.1 | - | 5 | 541 | ||
AK079752.1 | - | 5 | 662 | |||
BC016133.1 | - | 6 | 793 | 4485029 | ||
BC029056.1 | - | 5 | 582 | 4947517 |