Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144900.2 | NP_659149.1 | 23 | 3599 | ||
GenBank mRNA | AK088000.1 | - | 23 | 3560 | ||
AK137001.1 | - | 23 | 3559 | |||
AK148989.1 | - | 10 | 1457 | |||
AK150523.1 | - | 16 | 2505 | |||
AK159279.1 | - | 23 | 3556 | |||
AK159306.1 | - | 23 | 3554 | |||
BC010319.1 | - | 21 | 3184 | |||
BC021496.1 | - | 23 | 3549 | 5346159 | ||
BC023175.1 | - | 7 | 893 | |||
BC023794.1 | - | 24 | 3494 | 5351943 | ||
BC023867.1 | - | 6 | 895 | |||
BC023964.1 | - | 23 | 3540 | |||
BC025037.1 | - | 16 | 2399 | |||
BC025618.1 | - | 23 | 3558 | 5320976 | ||
BC025627.1 | - | 23 | 3549 | 5320513 | ||
BC025811.1 | - | 23 | 3536 | 5096872 | ||
BC030326.1 | - | 12 | 1896 | |||
BC031176.1 | - | 10 | 1457 | |||
BC031389.1 | - | 7 | 1018 | |||
BC032187.1 | - | 23 | 3599 | 5321268 | ||
BC033435.1 | - | 23 | 3495 | 4950746 | ||
BC033471.1 | - | 23 | 3539 | 4954119 | ||
BC034177.1 | - | 22 | 3304 | |||
BC035352.1 | - | 22 | 3269 | |||
BC042435.1 | - | 23 | 3549 | 4951288 |