Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 6330416G13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144905.3 | NP_659154.2 | 5 | 499 | ||
GenBank mRNA | AK030466.1 | - | 5 | 629 | ||
AK031016.1 | - | 5 | 503 | |||
AK138872.1 | - | 4 | 428 | |||
AK144990.1 | - | 6 | 676 | |||
AK161160.1 | - | 5 | 526 | |||
BC022599.1 | - | 3 | 353 | 4159235 | ||
BC023748.1 | - | 5 | 673 | 5353894 |