Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145073.2 | NP_659539.1 | 1 | 440 | ||
GenBank mRNA | BC125355.1 | - | 1 | 436 | 40129870 | |
BC125357.1 | - | 1 | 436 | 40129872 |