Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145153.3 | NP_660135.2 | 2 | 261 | ||
GenBank mRNA | AF481733.1 | - | 2 | 261 | ||
BC116433.2 | - | 2 | 261 | 40104472 | ||
BC116434.2 | - | 2 | 261 | 40104473 |