Gene Symbol: Mgat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145128.3 | NP_660110.2 | 4 | 951 | ||
GenBank mRNA | AF474154.1 | - | 2 | 331 | ||
AF474155.1 | - | 2 | 331 | |||
AK030402.1 | - | 4 | 919 | |||
AK036567.1 | - | 4 | 906 | |||
AK049587.1 | - | 3 | 873 | |||
BC125533.1 | - | 2 | 359 | 40130048 | ||
BC125535.1 | - | 2 | 359 | 40130050 |