Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smyd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144918.2 | NP_659167.2 | 11, 12 | 1069 | ||
GenBank mRNA | AK158267.1 | - | 11, 12 | 1050 | ||
AK162926.1 | - | 11, 12 | 993 | |||
BC016525.1 | - | 11, 12 | 1056 | 3601356 |