Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pitrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145131.1 | NP_660113.1 | 20 | 2266 | ||
GenBank mRNA | AF513714.1 | - | 20 | 2266 | ||
AK129292.1 | - | 20 | 2240 | |||
AK141277.1 | - | 20 | 2261 | |||
AK147837.1 | - | 20 | 2257 | |||
AK164235.1 | - | 20 | 2228 | |||
AK167426.1 | - | 20 | 2257 | |||
AK168335.1 | - | 19 | 2143 | |||
AY779273.1 | - | 20 | 2266 | |||
AY779274.1 | - | 20 | 2266 | |||
BC006917.1 | - | 10 | 1067 | 3601081 |