Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000818.2 | NP_000809.1 | 4 | 982 | ||
NM_001134366.1 | NP_001127838.1 | 4 | 982 | |||
GenBank mRNA | AK127375.1 | - | 4 | 1432 | ||
BC029517.1 | - | 4 | 556 | |||
BC126327.1 | - | 4 | 504 | 8992043 | ||
BC126329.1 | - | 4 | 504 | 8992045 | ||
M74826.1 | - | 4 | 620 | |||
M81882.1 | - | 4 | 531 | |||
X69936.1 | - | 4 | 524 |