Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TXNDC5 SIRNA Assays ›
Gene Symbol:
Txndc5
Gene Name:
thioredoxin domain containing 5
Gene Aliases:
AL022641, ERp46, PC-TRP
Chromosome Location:
Chr.13: 38500266 - 38528824 on Build GRCm38
UniGene ID:
Mm.28622
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
89288
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Txndc5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289598.1
NP_001276527.1
3
474
NM_001289599.1
NP_001276528.1
3
441
NM_145367.4
NP_663342.3
3
498
GenBank mRNA
AK136046.1
-
3
386
AK142049.1
-
3
484
AK150781.1
-
3
482
AK169680.1
-
3
465
AK224911.1
-
2
151
AK224913.1
-
3
485
AK224922.1
-
3
443
AK224923.1
-
3
465
AK224924.1
-
3
473
AK224926.1
-
3
443
AK224927.1
-
3
494
AK224930.1
-
3
446
AK224933.1
-
3
435
AK224934.1
-
3
489
AK224936.1
-
3
488
AK224937.1
-
3
487
AK224938.1
-
3
482
AK224939.1
-
3
475
AK224943.1
-
3
446
AK224945.1
-
3
482
AK224946.1
-
3
485
AK224947.1
-
3
472
AK224948.1
-
3
446
AK224949.1
-
3
435
AK224950.1
-
3
435
AK224951.1
-
3
493
AK224952.1
-
3
461
AK224956.1
-
3
406
AK224957.1
-
3
451
AK224958.1
-
3
451
AK224960.1
-
3
493
AK224962.1
-
3
453
AK224963.1
-
3
494
AK224964.1
-
3
494
AK224965.1
-
3
470
AK224966.1
-
3
470
AK224967.1
-
3
461
AK224968.1
-
1
28
AK224969.1
-
1
28
AK224970.1
-
3
435
AK224971.1
-
3
495
AK224972.1
-
3
485
AK224973.1
-
3
435
AK224974.1
-
3
438
AK224976.1
-
3
468
AK224977.1
-
3
482
AK224978.1
-
3
482
AK224979.1
-
3
469
AK224981.1
-
3
439
AK224982.1
-
3
494
AK224983.1
-
3
483
AK224984.1
-
3
448
AK224985.1
-
3
452
AK224986.1
-
3
496
AK224988.1
-
3
472
AK224989.1
-
3
489
AK224990.1
-
3
484
AK224991.1
-
3
479
AK224992.1
-
3
479
AK224993.1
-
3
479
AK224994.1
-
3
487
AK224995.1
-
3
450
AK224996.1
-
3
421
AK224997.1
-
3
446
AK224998.1
-
3
444
AK224999.1
-
3
466
AK225004.1
-
3
487
AK225005.1
-
3
465
AK225007.1
-
3
487
AK225010.1
-
3
487
AK225011.1
-
3
487
AK225012.1
-
3
256
AK225013.1
-
3
256
AK225014.1
-
3
465
AK225015.1
-
3
256
AK225016.1
-
3
395
AK225017.1
-
3
443
AK225019.1
-
3
475
AK225020.1
-
3
426
AK225021.1
-
3
426
AK225022.1
-
3
274
AK225023.1
-
3
500
AY243534.1
-
3
461
AY548112.1
-
3
422
BC016252.1
-
3
465
4911455
BC024505.1
-
3
410
4983496
BC036155.1
-
2
84
BC046789.1
-
3
424
5716897
BP765107.1
-
3
488
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001271330, FM045749, FM067602.
Panther Classification:
Molecular Function -
chaperone
Gene Ontology Categories:
Function(s)
Process(es)
protein folding
response to endoplasmic reticulum stress
apoptotic cell clearance
cell redox homeostasis
protein disulfide isomerase activity
isomerase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qskjf:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0