Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000856.5 | NP_000847.2 | 4 | 611 | ||
NM_001130682.2 | NP_001124154.1 | 3 | 517 | |||
NM_001130683.3 | NP_001124155.1 | 3 | 485 | |||
NM_001130684.2 | NP_001124156.1 | 2 | 457 | |||
NM_001130687.2 | NP_001124159.1 | 3 | 517 | |||
NM_001256449.1 | NP_001243378.1 | 3 | 448 | |||
XM_005262955.2 | XP_005263012.1 | 3 | 479 | |||
XM_006714196.2 | XP_006714259.1 | 3 | 466 | |||
XM_006714197.2 | XP_006714260.1 | 3 | 241 | |||
GenBank mRNA | AK096588.1 | - | 3 | 448 | ||
AK309950.1 | - | 3 | 379 | |||
AK312952.1 | - | 3 | 417 | |||
BC012627.1 | - | 2 | 273 | |||
BC028384.2 | - | 2 | 457 | 4792661 | ||
BC095395.1 | - | 3 | 408 | |||
BX647200.1 | - | 3 | 284 | |||
BX649180.1 | - | 3 | 378 | |||
DA518091.1 | - | 3 | 486 | |||
DA793192.1 | - | 3 | 517 | |||
DA908917.1 | - | 3 | 485 | |||
U58855.1 | - | 1 | 92 | |||
X66534.1 | - | 4 | 597 | |||
Y15723.1 | - | 4 | 598 |