Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145405.2 | NP_663380.1 | 2, 3 | 237 | ||
GenBank mRNA | AJ627038.1 | - | 2, 3 | 198 | ||
AK146110.1 | - | 227 | ||||
BC010817.1 | - | 2, 3 | 216 | 4215699 | ||
BM932729.1 | - | 2, 3 | 211 |
Gene Symbol: Gm38419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278271.1 | NP_001265200.1 | 5, 6 | 836 | ||
NM_001278272.1 | NP_001265201.1 | 5, 6 | 800 | |||
GenBank mRNA | EU599043.1 | - | 5, 6 | 705 | ||
EU599044.1 | - | 5, 6 | 669 |