Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Txndc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289598.1 | NP_001276527.1 | 10 | 1585 | ||
NM_001289599.1 | NP_001276528.1 | 10 | 1552 | |||
NM_145367.4 | NP_663342.3 | 10 | 1609 | |||
GenBank mRNA | AK142049.1 | - | 10 | 1595 | ||
AK150781.1 | - | 10 | 1593 | |||
AK169680.1 | - | 10 | 1576 | |||
AK224911.1 | - | 9 | 1262 | |||
AK224912.1 | - | 1 | 60 | |||
AK224913.1 | - | 10 | 1596 | |||
AK224922.1 | - | 10 | 1554 | |||
AK224923.1 | - | 10 | 1576 | |||
AK224924.1 | - | 10 | 1584 | |||
AK224926.1 | - | 10 | 1554 | |||
AK224927.1 | - | 10 | 1605 | |||
AK224930.1 | - | 10 | 1557 | |||
AK224933.1 | - | 10 | 1546 | |||
AK224934.1 | - | 10 | 1600 | |||
AK224935.1 | - | 3 | 1751 | |||
AK224936.1 | - | 10 | 1599 | |||
AK224937.1 | - | 10 | 1598 | |||
AK224938.1 | - | 10 | 1593 | |||
AK224939.1 | - | 10 | 1586 | |||
AK224943.1 | - | 10 | 1557 | |||
AK224945.1 | - | 10 | 1593 | |||
AK224946.1 | - | 10 | 1596 | |||
AK224947.1 | - | 10 | 1583 | |||
AK224948.1 | - | 10 | 1557 | |||
AK224949.1 | - | 10 | 1546 | |||
AK224950.1 | - | 10 | 1546 | |||
AK224951.1 | - | 10 | 1604 | |||
AK224953.1 | - | 1 | 202 | |||
AK224955.1 | - | 5 | 857 | |||
AK224956.1 | - | 10 | 1516 | |||
AK224957.1 | - | 10 | 1562 | |||
AK224958.1 | - | 10 | 1562 | |||
AK224960.1 | - | 10 | 1604 | |||
AK224962.1 | - | 10 | 1564 | |||
AK224967.1 | - | 10 | 1572 | |||
AK224968.1 | - | 8 | 1139 | |||
AK224969.1 | - | 8 | 1139 | |||
AK224970.1 | - | 10 | 1546 | |||
AK224971.1 | - | 10 | 1606 | |||
AK224972.1 | - | 10 | 1596 | |||
AK224973.1 | - | 10 | 1546 | |||
AK224974.1 | - | 10 | 1549 | |||
AK224975.1 | - | 5 | 784 | |||
AK224976.1 | - | 10 | 1579 | |||
AK224977.1 | - | 10 | 1593 | |||
AK224978.1 | - | 10 | 1593 | |||
AK224979.1 | - | 10 | 1580 | |||
AK224981.1 | - | 10 | 1550 | |||
AK224982.1 | - | 10 | 1605 | |||
AK224983.1 | - | 10 | 1594 | |||
AK224984.1 | - | 10 | 1559 | |||
AK224985.1 | - | 10 | 1563 | |||
AK224986.1 | - | 10 | 1606 | |||
AK224988.1 | - | 10 | 1583 | |||
AK224989.1 | - | 10 | 1600 | |||
AK224990.1 | - | 10 | 1595 | |||
AK224991.1 | - | 10 | 1590 | |||
AK224992.1 | - | 10 | 1590 | |||
AK224993.1 | - | 10 | 1590 | |||
AK224994.1 | - | 10 | 1598 | |||
AK224995.1 | - | 10 | 1561 | |||
AK224996.1 | - | 10 | 1532 | |||
AK224997.1 | - | 10 | 1557 | |||
AK224998.1 | - | 10 | 1555 | |||
AK224999.1 | - | 10 | 1577 | |||
AK225000.1 | - | 1 | 370 | |||
AK225004.1 | - | 10 | 1598 | |||
AK225006.1 | - | 1 | 53 | |||
AK225007.1 | - | 10 | 1598 | |||
AK225008.1 | - | 1 | 53 | |||
AK225010.1 | - | 10 | 1598 | |||
AK225011.1 | - | 10 | 1598 | |||
AK225012.1 | - | 10 | 1367 | |||
AK225013.1 | - | 10 | 1367 | |||
AK225014.1 | - | 10 | 1576 | |||
AK225015.1 | - | 10 | 1367 | |||
AK225016.1 | - | 10 | 1506 | |||
AK225017.1 | - | 10 | 1554 | |||
AK225019.1 | - | 10 | 1586 | |||
AK225020.1 | - | 10 | 1537 | |||
AK225021.1 | - | 10 | 1537 | |||
AK225022.1 | - | 10 | 1385 | |||
AK225023.1 | - | 10 | 1611 | |||
AY243534.1 | - | 10 | 1572 | |||
BC016252.1 | - | 10 | 1576 | 4911455 | ||
BC024505.1 | - | 10 | 1521 | 4983496 | ||
BC036155.1 | - | 9 | 1195 | |||
BC046789.1 | - | 10 | 1535 | 5716897 |