Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mfsd7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145447.2 | NP_663422.1 | 10 | 2532 | ||
GenBank mRNA | AK049494.1 | - | 1 | 49 | ||
AK145696.1 | - | 10 | 2488 | |||
AY260575.1 | - | 10 | 2532 | |||
AY260576.1 | - | 10 | 2532 | |||
BC011209.1 | - | 10 | 2532 | 4190027 |