Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039530.3 | NP_001034619.2 | 14 | 4426 | ||
GenBank mRNA | AK220263.1 | - | 9 | 2018 | ||
BC021340.1 | - | 2 | 277 | |||
BC047386.1 | - | 9 | 1526 | |||
BC138874.1 | - | 14 | 4541 | 8861896 | ||
BC145452.1 | - | 13 | 4628 | |||
DQ372930.1 | - | 14 | 4590 |