Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145462.2 | NP_663437.2 | 4 | 453 | ||
GenBank mRNA | AK079161.1 | - | 4 | 393 | ||
AK079852.1 | - | 4 | 421 | |||
AK087969.1 | - | 4 | 428 | |||
AK196117.1 | - | 2 | 98 | |||
AK216105.1 | - | 2 | 97 | |||
BB855694.1 | - | 4 | 454 | |||
BC004644.1 | - | 4 | 428 | 3157901 | ||
BC011095.1 | - | 4 | 428 | 3157901 |