Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sft2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145512.4 | NP_663487.1 | 6 | 502 | ||
GenBank mRNA | AK036578.1 | - | 6 | 390 | ||
AK075812.1 | - | 5 | 332 | |||
AK156973.1 | - | 6 | 393 | |||
AK170590.1 | - | 5 | 307 | |||
BC017549.1 | - | 6 | 381 | 4924681 |