Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079849.2 | NP_001073318.1 | 10 | 1074 | ||
NM_145457.4 | NP_663432.1 | 10 | 1292 | |||
GenBank mRNA | AF128835.1 | - | 9 | 974 | ||
BC019726.1 | - | 10 | 1263 | 4010170 | ||
BC051042.1 | - | 10 | 1249 | 4008643 |