Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145488.1 | NP_663463.1 | 11 | 2193 | ||
GenBank mRNA | AK143173.1 | - | 11 | 2181 | ||
AK207198.1 | - | 1 | 36 | |||
AK220525.1 | - | 11 | 2174 | |||
BC003424.1 | - | 11 | 2193 | 2609884 |