Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lzts2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130525.1 | NP_001123997.1 | 5 | 2587 | ||
NM_001130526.1 | NP_001123998.1 | 5 | 2579 | |||
NM_145503.2 | NP_663478.2 | 5 | 2631 | |||
GenBank mRNA | AK081367.1 | - | 5 | 2506 | ||
AK129446.1 | - | 4 | 2496 | |||
AK144154.1 | - | 5 | 2598 | |||
AK202462.1 | - | 1 | 82 | |||
AK205448.1 | - | 1 | 82 | |||
AK210216.1 | - | 1 | 82 | |||
AK215584.1 | - | 1 | 82 | |||
AK215776.1 | - | 1 | 82 | |||
AK217660.1 | - | 1 | 82 | |||
AK217765.1 | - | 1 | 24 | |||
AK218561.1 | - | 1 | 82 | |||
BC014695.1 | - | 4 | 2480 | 3157493 | ||
BC092263.1 | - | 5 | 2611 | 2654420 |