Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145516.2 | NP_663491.1 | 8 | 1739 | ||
GenBank mRNA | AF189817.1 | - | 8 | 1719 | ||
AK039771.1 | - | 8 | 1707 | |||
AK166091.1 | - | 8 | 1707 | |||
BC057994.1 | - | 10 | 1098 | 5344583 |