Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145582.1 | NP_663557.1 | 2 | 193 | ||
GenBank mRNA | AK155284.1 | - | 2 | 208 | ||
AK160967.1 | - | 2 | 194 | |||
BC005752.1 | - | 2 | 193 | 2654430 |