Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem184c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145599.4 | NP_663574.3 | 1, 2 | 559 | ||
GenBank mRNA | AK033404.1 | - | 1, 2 | 528 | ||
AK164182.1 | - | 1, 2 | 527 | |||
BC004056.1 | - | 1, 2 | 517 | 3589819 | ||
CN532034.1 | - | 1, 2 | 559 |