Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtnr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145712.2 | NP_663758.2 | 1, 2 | 327 | ||
GenBank mRNA | AB377276.1 | - | 327 | |||
AY145850.1 | - | 1, 2 | 231 | |||
BC104324.1 | - | 1, 2 | 231 | 40044921 | ||
BC104325.1 | - | 1, 2 | 231 | 40044928 |
Set Membership: |
G-protein coupled receptor |