Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp772 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145577.2 | NP_663552.2 | 2, 3 | 328 | ||
GenBank mRNA | AK142307.1 | - | 2, 3 | 328 | ||
BC023179.1 | - | 2, 3 | 284 | 4951074 | ||
BC108353.1 | - | 2, 3 | 219 | 5067589 |