Search Thermo Fisher Scientific
Gene Symbol: Fgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145594.2 | NP_663569.2 | 7 | 751 | ||
GenBank mRNA | AF478470.1 | - | 7 | 761 | ||
BC021946.1 | - | 7 | 719 | 5098805 | ||
BC029734.1 | - | 7 | 726 | 4158843 |