Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145630.2 | NP_663605.1 | 11 | 1341 | ||
GenBank mRNA | AK075879.1 | - | 11 | 1337 | ||
AK075985.1 | - | 11 | 1397 | |||
AK088478.1 | - | 11 | 1422 | |||
AK132024.1 | - | 11 | 1365 | |||
AK169874.1 | - | 11 | 1422 | |||
BC008126.1 | - | 11 | 1341 | 3500763 | ||
CT010338.1 | - | 11 | 1198 |