Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000873.3 | NP_000864.2 | 2 | 456 | ||
NM_001099786.1 | NP_001093256.1 | 4 | 579 | |||
NM_001099787.1 | NP_001093257.1 | 3 | 497 | |||
NM_001099788.1 | NP_001093258.1 | 4 | 462 | |||
NM_001099789.1 | NP_001093259.1 | 3 | 380 | |||
GenBank mRNA | AB209878.1 | - | 3 | 488 | ||
AF340052.1 | - | 2 | 261 | |||
AF340053.1 | - | 2 | 270 | |||
AK223219.1 | - | 2 | 300 | |||
AK293110.1 | - | 2 | 302 | |||
AK295382.1 | - | 3 | 341 | |||
BC003097.1 | - | 2 | 278 | 3502827 | ||
BM695682.1 | - | 2 | 456 | |||
BX363874.2 | - | 4 | 524 | |||
CR541812.1 | - | 2 | 224 | |||
CR541834.1 | - | 2 | 224 | |||
X15606.1 | - | 2 | 286 |