Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D15Ertd621e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145959.3 | NP_666071.2 | 24 | 4565 | ||
GenBank mRNA | AK122297.1 | - | 24 | 4415 | ||
AK137324.1 | - | 24 | 4567 | |||
BC033609.1 | - | 16 | 3647 | 5401763 | ||
BC070426.1 | - | 24 | 4551 | 6405871 |