Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146061.4 | NP_666173.4 | 2 | 382 | ||
GenBank mRNA | AF482997.1 | - | 2 | 377 | ||
AK046057.1 | - | 3 | 199 | |||
AK081231.1 | - | 2 | 378 | |||
BC024991.1 | - | 3 | 266 | 5065129 |