Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apbb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146085.1 | NP_666197.1 | 2 | 456 | ||
GenBank mRNA | AK141878.1 | - | 2 | 358 | ||
AK171634.1 | - | 2 | 407 | |||
BC024136.1 | - | 2 | 212 | |||
BC024809.1 | - | 2 | 456 | 5357681 |