Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spidr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146068.4 | NP_666180.2 | 3 | 293 | ||
GenBank mRNA | AK044836.1 | - | 3 | 276 | ||
AK052832.1 | - | 3 | 258 | |||
AK075871.1 | - | 3 | 258 | |||
AK146053.1 | - | 3 | 293 | |||
AK172902.1 | - | 3 | 246 | |||
BC089026.1 | - | 3 | 230 | 30615298 |