Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146093.1 | NP_666205.1 | 6 | 401 | ||
GenBank mRNA | AK015468.1 | - | 5 | 453 | ||
AK150442.1 | - | 5 | 404 | |||
AK159708.1 | - | 5 | 377 | |||
AK172428.1 | - | 3 | 755 | |||
BC006701.1 | - | 6 | 401 | 3583916 |