Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271356.1 | NP_001258285.1 | 12, 13 | 2010 | ||
NM_001271357.1 | NP_001258286.1 | 12, 13 | 1753 | |||
NM_146018.2 | NP_666130.1 | 12, 13 | 2050 | |||
GenBank mRNA | AK039106.1 | - | 12, 13 | 1837 | ||
AK080933.1 | - | 12, 13 | 1835 | |||
AK136071.1 | - | 12, 13 | 1741 | |||
AK148216.1 | - | 12, 13 | 1804 | |||
AK154040.1 | - | 12, 13 | 2032 | |||
AK161460.1 | - | 12, 13 | 2093 | |||
AK172245.1 | - | 12, 13 | 1808 | |||
BC025820.1 | - | 12, 13 | 1784 | 5099493 |