Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146061.4 | NP_666173.4 | 8 | 1274 | ||
GenBank mRNA | AF482997.1 | - | 8 | 1269 | ||
AK028831.1 | - | 1 | 393 | |||
AK046057.1 | - | 9 | 1092 | |||
AK081231.1 | - | 8 | 1270 | |||
BC024991.1 | - | 9 | 1158 | 5065129 | ||
BE653785.1 | - | 1 | 296 |