Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aph1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146104.3 | NP_666216.1 | 6 | 1605 | ||
NM_146134.2 | NP_666246.1 | 7 | 1272 | |||
GenBank mRNA | AK050828.1 | - | 4 | 1829 | ||
AK078343.1 | - | 4 | 1070 | |||
AK148400.1 | - | 6 | 1558 | |||
AK152334.1 | - | 6 | 1548 | |||
AK181631.1 | - | 2 | 328 | |||
AK217389.1 | - | 1 | 284 | |||
AX775953.1 | - | 6 | 1335 | |||
BC012406.1 | - | 6 | 1530 | 3483214 | ||
BC024111.1 | - | 7 | 1138 | 5098973 | ||
BC057865.1 | - | 6 | 1548 | 5320175 | ||
CX207467.1 | - | 3 | 460 |