Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbxip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146131.2 | NP_666243.1 | 3 | 205 | ||
GenBank mRNA | AK028754.1 | - | 3 | 166 | ||
AK028858.1 | - | 3 | 203 | |||
AK159422.1 | - | 3 | 192 | |||
AK159448.1 | - | 3 | 192 | |||
AK159633.1 | - | 3 | 182 | |||
AK160103.1 | - | 3 | 186 | |||
AK170318.1 | - | 3 | 161 | |||
BC026838.1 | - | 2 | 140 | 4460993 | ||
BC027771.1 | - | 3 | 175 | 5344900 | ||
BC034200.1 | - | 3 | 146 | 4986595 |