Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146175.3 | NP_666287.3 | 1, 2 | 290 | ||
GenBank mRNA | AK030516.1 | - | 1, 2 | 199 | ||
AK081854.1 | - | 1, 2 | 189 | |||
AK088531.1 | - | 1, 2 | 289 | |||
AK148038.1 | - | 1, 2 | 276 | |||
AK188102.1 | - | 1, 2 | 108 | |||
BC025581.1 | - | 1, 2 | 192 | 5325029 |