Search Thermo Fisher Scientific
Gene Symbol: Ppa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293641.1 | NP_001280570.1 | 2, 3 | 245 | ||
NM_146141.2 | NP_666253.1 | 2, 3 | 245 | |||
GenBank mRNA | AA444705.1 | - | 2, 3 | 244 | ||
AK090384.1 | - | 2, 3 | 234 | |||
AK143475.1 | - | 2, 3 | 216 | |||
BC011417.1 | - | 2, 3 | 239 | 2654305 |