Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146145.2 | NP_666257.2 | 8 | 1458 | ||
GenBank mRNA | AK133866.1 | - | 8 | 1401 | ||
AK141210.1 | - | 8 | 1400 | |||
AK143650.1 | - | 8 | 1404 | |||
AK148521.1 | - | 8 | 1422 | |||
AK152128.1 | - | 8 | 1421 | |||
S63728.1 | - | 8 | 1408 |