Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thrap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146153.3 | NP_666265.3 | 3 | 470 | ||
GenBank mRNA | AK034044.1 | - | 3 | 350 | ||
AK050208.1 | - | 1 | 1986 | |||
AK079601.1 | - | 3 | 350 | |||
BC026580.1 | - | 3 | 372 | |||
BC040346.1 | - | 3 | 401 | |||
BC092239.1 | - | 3 | 386 | 30659321 |