Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Picalm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252520.1 | NP_001239449.1 | 21 | 3171 | ||
NM_001252521.1 | NP_001239450.1 | 20 | 3162 | |||
NM_001252522.1 | NP_001239451.1 | 20 | 3036 | |||
NM_001252523.1 | NP_001239452.1 | 20 | 3021 | |||
NM_001252524.1 | NP_001239453.1 | 19 | 2997 | |||
NM_146194.4 | NP_666306.2 | 21 | 3186 | |||
GenBank mRNA | AK085472.2 | - | 16 | 2231 | ||
AK144960.1 | - | 21 | 2950 | |||
AK162360.1 | - | 21 | 2864 | |||
AK165066.1 | - | 15 | 2134 | |||
AK170373.1 | - | 19 | 3017 | |||
AK220291.1 | - | 21 | 2887 | |||
BC011470.1 | - | 21 | 2881 | 2651109 | ||
BC021491.1 | - | 19 | 2946 | 5345558 | ||
BC023843.1 | - | 20 | 2910 | 5346041 | ||
BC025566.1 | - | 20 | 3059 | 5322606 | ||
BC027116.1 | - | 17 | 2458 |