Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbxip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146131.2 | NP_666243.1 | 9, 10 | 989 | ||
GenBank mRNA | AK028754.1 | - | 9, 10 | 950 | ||
AK028858.1 | - | 9, 10 | 987 | |||
AK159422.1 | - | 9, 10 | 976 | |||
AK159448.1 | - | 9, 10 | 976 | |||
AK159633.1 | - | 9, 10 | 966 | |||
AK160103.1 | - | 9, 10 | 970 | |||
AK170318.1 | - | 9, 10 | 945 | |||
BC026838.1 | - | 8, 9 | 924 | 4460993 | ||
BC027771.1 | - | 9, 10 | 959 | 5344900 | ||
BC034200.1 | - | 9, 10 | 930 | 4986595 |