Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cul4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146207.2 | NP_666319.2 | 20 | 2247 | ||
GenBank mRNA | AK003679.1 | - | 4 | 448 | ||
AK005426.1 | - | 5 | 550 | |||
AK144802.1 | - | 20 | 2230 | |||
AK168215.1 | - | 20 | 2229 | |||
AK170722.1 | - | 20 | 2229 | |||
AK190734.1 | - | 3 | 301 | |||
AK194132.1 | - | 3 | 329 | |||
BC010211.1 | - | 20 | 2225 | 3969515 | ||
BC024113.1 | - | 16 | 1719 | 5097944 | ||
BC024667.1 | - | 2 | 233 | |||
BC027006.1 | - | 16 | 1767 |